Learn More
MP Biomedicals™ Geneclean Glassmilk
Catalog No. 111001204 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Packaging Quantity:
1.25 mL
Catalog No. Packaging Quantity
11-100-1204 1.25 mL
1 options
Catalog No. 11-100-1204 Supplier MP Biomedicals™ Supplier No. 111001204
Add to Cart
Add to Cart
GeneClean Glassmilk
  • Glassmilk slurry allows flexibility of reaction size and volume.
  • GeneClean Glassmilk is a specially prepared aqueous suspension of proprietary silica matrix.
  • GeneClean Glassmilk is supplied with GeneClean Kit, which will bind to radioactive isotope-labeled ssDNA or dsDNA that is purified from agarose thus resulting in purified DNA for further use.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Kits and Assays
Description Geneclean™glassmilk™ is a specially prepared aqueous suspension of proprietary silica matrix.
Packaging Quantity 1.25 mL
WARNING: Cancer - www.P65Warnings.ca.gov
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.