Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ GeneClean II Kit

Catalog No. 111001400 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
300 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
11-100-1400 300 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-1400 Supplier MP Biomedicals™ Supplier No. 111001400
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GeneClean™ II Kit

  • The GENECLEAN™ II Kit is designed to optimally purify 200 bp to 20 kb DNA from TAE and TBE buffered agarose gels and solutions. Glassmilk™ slurry allows flexibility of reacion size and volume.
  • The purified DNA is suitable for a variety of downstream applications, such as sequencing, PCR, restriction digestion, cloning and labeling.
  • The GENECLEAN™ II Kit is designed for the rapid purification of linear and plasmid DNA fragments from standard or low-melting agarose gels.
  • This kit can also be used to purify DNA from polyacrylamide gels.
  • Typical recovery of DNA is up to 80%.
  • Each column can bind up to 10 μg of DNA from up to a 3.5 g agarose slice.
  • The GENECLEAN™ II Kit combines silica-binding technology with the convenience of a spin or vacuum column format.
  • DNA fragments of interest are extracted from slices of an agarose gel and are bound to a silica membrane.
  • Contaminants are removed by a simple spin or vacuum wash.
  • The bound DNA is then eluted.
  • DNA fragments of interest are extracted by solubilizing slices of an agarose gel.
  • The Gel Solubilization Solution dissolves an agarose gel slice from gels run in TBE or TAE buffer.
  • The extracted DNA fragments selectively adsorb onto the silica membrane in the presence of the Gel Solubilization Solution.
  • Contaminants are removed by a simple spin or vacuum wash. Finally, the bound DNA is eluted in Tris buffer.
TRUSTED_SUSTAINABILITY

Specifications

Concentration New Wash/Ethanol Concentration must be ∽ 50%
Content And Storage All gENECLEAN™ Kits are shipped at room temperature (15-30°C).
Format Kit
Isolation Technology Silica Matrix Technology
For Use With (Application) Nucleic Acid Isolation & Purification
No. of Reactions 300 Preps
pH Above 500 bp: 4.5 M NaI pH 7.0 - 7.4; Between 200 - 500 bp: 4.5 M NaI pH 6.0; NEW Wash: pH Range: 7.4 - 8.0
Product Type Kits & Assays
Synonym Rapid purification of linear DNA
Form Solution or Gels
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.