Learn More
Agilent Technologies GENEJAMMER TRANSFECT REAGENT
Catalog No. 50125202
Encompass
GeneJammer Transfection Reagent 1mL 1.33 mg/mL reagent in 80% ethanol Applications Incorporation of foreign genes into mammalian cells including primary cultures and hard-to-transfect cell lines Transfects DNA
Catalog No. 50-125-202 Supplier Agilent Technologies Supplier No. 204130
GeneJammer Transfection Reagent 1mL 1.33 mg/mL reagent in 80% ethanol Applications Incorporation of foreign genes into mammalian cells including primary cultures and hard-to-transfect cell lines Transfects DNA