Learn More
Agilent Technologies GENEJAMMER TRANSFECT REAGENT
Supplier:  Agilent Technologies 204132
Encompass
GeneJammer Transfection Reagent Sampler 0.3mL 1.33 mg/mL reagent in 80% ethanol Applications Incorporation of foreign genes into mammalian cells including primary cultures and hard-to-transfect cell lines Transfects DNA
Catalog No. 50-125-204