Learn More
Agilent Technologies GENEJAMMER TRANSFECT REAGENT
Supplier:  Agilent Technologies 204131
Encompass
GeneJammer Transfection Reagent is a proprietary formulation of polyamine and other components in 80% ethanol ideal for transient or stable transfections in a wide variety of cell lines .Fast, easy to use with low cytotoxicity
Catalog No. 50-125-203