Learn More
Sigma Aldrich Fine Chemicals Biosciences GenEluteTM Bacterial Genom
Catalog No. 502901887
Encompass
The GenElute Bacterial Genomic DNA kit provides a simple and convenient way to isolate pure genomic DNA from gram-negative bacteria. for most gram-positive bacteria the kit must be used in conjunction with the optional lysozyme (L4919) to effectively lyse the thick peptidoglycan cell walls. A Gram-Positive Lysis SOLUTIONon is provided as a diluent for preparing the lysozyme stock SOLUTIONons.The kit combines the advantages of a silica-based system with a microspin format and eliminates the need for expensive resins alcohol precipitation and hazardous organic compounds such as phenol and chloroform.
Catalog No. 50-290-1887 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. NA21101KT
The GenElute Bacterial Genomic DNA kit provides a simple and convenient way to isolate pure genomic DNA from gram-negative bacteria. for most gram-positive bacteria the kit must be used in conjunction with the optional lysozyme (L4919) to effectively lyse the thick peptidoglycan cell walls. A Gram-Positive Lysis SOLUTIONon is provided as a diluent for preparing the lysozyme stock SOLUTIONons.The kit combines the advantages of a silica-based system with a microspin format and eliminates the need for expensive resins alcohol precipitation and hazardous organic compounds such as phenol and chloroform.