Learn More
Sigma Aldrich Fine Chemicals Biosciences GenEluteTM Blood Genomic D
Catalog No. 502907270
Encompass
The GenElute Blood Genomic DNA kit provides a simple and convenient way to isolate pure genomic DNA from fresh or aged (older than 24 hours) whole blood. The kit combines the advantages of silica binding with a microspin format and eliminates the need for expensive resins alcohol precipitation and hazardous organic compounds such as phenol and chloroform.
Catalog No. 50-290-7270 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. NA20201KT
The GenElute Blood Genomic DNA kit provides a simple and convenient way to isolate pure genomic DNA from fresh or aged (older than 24 hours) whole blood. The kit combines the advantages of silica binding with a microspin format and eliminates the need for expensive resins alcohol precipitation and hazardous organic compounds such as phenol and chloroform.