Learn More
Agilent Technologies GENEMORPH II RANDOM MUTA KIT
Catalog No. NC9107220
Encompass
GeneMorph II Random Mutagenesis Kit, Contains: Mutazyme II DNA Polymerase 10X Mutazyme Reaction Buffer 40mM dNTP mix 1.1 kb Gel Standard, 30 rxn, 1/KT
Catalog No. NC9107220 Supplier Agilent Technologies Supplier No. 200550
GeneMorph II Random Mutagenesis Kit, Contains: Mutazyme II DNA Polymerase 10X Mutazyme Reaction Buffer 40mM dNTP mix 1.1 kb Gel Standard, 30 rxn, 1/KT