Learn More
Amsbio LLC GenePORTER Gold Transfection Reagent
Catalog No. 501632249
Encompass
The GenePORTER Gold Transfection Reagent is our newest, most effective and reproducible lipid based gene delivery formulation for a wide variety of cell lines. It utilizes our Advanced Carrier Enhancement (ACE) technology that maximizes transfection efficiency and transgene expression levels with minimal cytotoxicity. The transfection efficiency levels achieved with GenePORTER Gold match or surpass other commercially available reagents. The GenePORTER Gold kit features an easy to use protocol and enough reagent for 400 x 1ug transfections to ensure that you get maximum transfection efficiency and value per reaction in the broadest spectrum of cell lines.
Catalog No. 50-163-2249 Supplier Amsbio LLC Supplier No. AMS.T204030
The GenePORTER Gold Transfection Reagent is our newest, most effective and reproducible lipid based gene delivery formulation for a wide variety of cell lines. It utilizes our Advanced Carrier Enhancement (ACE) technology that maximizes transfection efficiency and transgene expression levels with minimal cytotoxicity. The transfection efficiency levels achieved with GenePORTER Gold match or surpass other commercially available reagents. The GenePORTER Gold kit features an easy to use protocol and enough reagent for 400 x 1ug transfections to ensure that you get maximum transfection efficiency and value per reaction in the broadest spectrum of cell lines.