Learn More
Alpco Ltd GeneProof Aspergillus PCR Kit, 100 tests
SDP
Catalog No. NC2271527
Encompass
The GeneProof Aspergillus PCR Kit boasts high sensitivity by targeting the multicopy sequence of the Intergenic spacer ITS2/28S rDNA. For Research Use Only in North America. This kit is sufficient for 100 tests.
This item is not returnable. View return policy
Catalog No. NC2271527 Supplier Alpco Ltd Supplier No. 60ASP002100
The GeneProof Aspergillus PCR Kit boasts high sensitivity by targeting the multicopy sequence of the Intergenic spacer ITS2/28S rDNA. For Research Use Only in North America. This kit is sufficient for 100 tests.