Learn More
Alpco Ltd GeneProof Epstein-Barr Virus (EBV) PCR Kit
SDP
Catalog No. NC2271640
Encompass
The GeneProof Epstein-Barr Virus (EBV) PCR Kit is precise and fully traceable quantification according to WHO 1st International Standard NIBSC 09/260. For Research Use Only in North America. This kit is sufficient for 100 tests.
This item is not returnable. View return policy
Catalog No. NC2271640 Supplier Alpco Ltd Supplier No. 60EBV002100
The GeneProof Epstein-Barr Virus (EBV) PCR Kit is precise and fully traceable quantification according to WHO 1st International Standard NIBSC 09/260. For Research Use Only in North America. This kit is sufficient for 100 tests.