Learn More
Alpco Ltd GeneProof Parvovirus B19 PCR Kit
SDP
Supplier:  Alpco Ltd 60B19002100
Encompass
The Parvovirus B19 IgG Enzyme Immunoassay Kit provides materials for the qualitative and semiquantitative determination of IgG-class antibodies to Parvovirus B19 in human serum. Research Use Only. Not for Use in Diagnostic Procedures. This kit is sufficient for 100 tests.
Catalog No. NC2284570