Learn More
Alpco Ltd GeneProof Parvovirus B19 PCR Kit
SDP
Catalog No. NC2284569
Encompass
The Parvovirus B19 IgG Enzyme Immunoassay Kit provides materials for the qualitative and semiquantitative determination of IgG-class antibodies to Parvovirus B19 in human serum. Research Use Only. Not for Use in Diagnostic Procedures. This kit is sufficient for 25 tests.
This item is not returnable. View return policy
Catalog No. NC2284569 Supplier Alpco Ltd Supplier No. 60B19002025
The Parvovirus B19 IgG Enzyme Immunoassay Kit provides materials for the qualitative and semiquantitative determination of IgG-class antibodies to Parvovirus B19 in human serum. Research Use Only. Not for Use in Diagnostic Procedures. This kit is sufficient for 25 tests.