Learn More
Alpco Ltd GeneProof SARS-CoV-2 Screening PCR Kit
SDP
Catalog No. NC2284571
Encompass
The GeneProof SARS-CoV-2 Screening PCR Kit contains three targets (RdRp/E/N genes) in one reaction is a robust and reliable screening solution for two-channel thermocyclers. For Research Use Only In North America. This kit is sufficient for 25 tests.
This item is not returnable. View return policy
Catalog No. NC2284571 Supplier Alpco Ltd Supplier No. 60COV2S002025
The GeneProof SARS-CoV-2 Screening PCR Kit contains three targets (RdRp/E/N genes) in one reaction is a robust and reliable screening solution for two-channel thermocyclers. For Research Use Only In North America. This kit is sufficient for 25 tests.