Learn More
Alpco Ltd GeneProof Treponema pallidum PCR Kit
SDP
Supplier:  Alpco Ltd 60TP002025
Encompass
The GeneProof Treponema pallidum PCR Kit targets the highly conservative polA gene for DNA polymerase I. For Research Use Only in North America. This kit is sufficient for 25 tests.
Catalog No. NC2284579