Learn More
Corning™ General Assay Microplate Lids
GSA_VAGREENER_CHOICE
Catalog No. 07200703 Shop All Corning Products
Change view
Click to view available options
Plate Compatibility:
96-well Microplates Only (not 2 mL block)
96/384-well, Half Area Plate
All Microplates
No. per Case:
100/Cs.
50/Cs.
Material:
Polystyrene
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plate Compatibility No. per Case Material
07-200-703 96/384-well, Half Area Plate 50/Cs. Polystyrene
07-200-598 96-well Microplates Only (not 2 mL block) 50/Cs. Polystyrene
07-200-694 All Microplates 100/Cs. Polystyrene
07-200-597 96-well Microplates Only (not 2 mL block) 100/Cs. Polystyrene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 07-200-703 Supplier Corning™ Supplier No. 3099
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Manufactured with rigid polystyrene for optimum visibility and strength. Corning™ General Assay Microplate Lids are available in a variety of styles and types.
  • All lids are made of rigid polystyrene except where indicated.
  • All lids have a corner notch on the A1 corner (except where indicated) to correspond to the corner notches found on all Corning™ microplates.
  • The universal lid without a corner notch (Cat. No. 3098) does not need to be oriented in any particular direction to be placed on Corning microplates. The lid also has a shorter skirt than standard lids.
  • The black universal lid (Cat. No. 3935) is suitable for fluorescent and other light-sensitive assays.
  • The DMSO-resistant cyclic olefin copolymer (COC) lid (Cat. No. 3085) is tinted amber in color for light-sensitive assays and is 100% DMSO-resistant.
  • This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
    TRUSTED_SUSTAINABILITY
    Specifications
    Material Polystyrene
    Sterility Sterile
    No. per Case 50/Cs.
    No. per Pack 25/Pk.
    Product Type Universal Lid
    Plate Compatibility 96/384-well, Half Area Plate
    Cut-off Corners Corner Notch