Learn More
Fisherbrand™ General Biology Dissecting Kit
Catalog No. 13820097 Shop All Fisherbrand Products
Change view
Click to view available options
Includes:
8 Pcs
Catalog No. Includes
13-820-097 8 Pcs
1 options
Catalog No. 13-820-097
Add to Cart
Add to Cart
The best dissecting kits for your needs!
Fisherbrand™ General Biology Dissecting Kit:
  • Single fold vinyl case w/ lining (Blue)
  • #4 surgical Handle w/ #21 replaceable Blade
  • 5-1/2 in. Sharp/Blunt operating Scissors
  • 5-1/2 in. Straight Thumb Forceps:
  • Round, Serr. Tips; Serr. Grip
  • Stainless steel mall-probe and seeker:
  • Hexagon 3¾ in. Handle; 2½ in. Seeker
  • Straight teasing Needle w/ 4 in.Plastic Handle
  • Glass dropper w/ rubber bulb- large Size 5 in.
  • 6 in. hard Plastic ruler
Certifications
ISO 9001-02, GMP, cGMP
Specifications
Product Type Dissection Tool Kit
For Use With (Application) Dissecting Experiments
Includes 8 Pcs
Quantity 8 Pc.