Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific General Chemistry Calibrators

Catalog No. 23666479 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
1 x 2 mL
6 x 1 mL
10 x 5 mL
Type:
AutoHDL™/LDL Calibrator
Chemistry Calibrator
Lipoprotein A Calibrator Set
Microalbumin Calibrator Set
Color:
Clear
Light Yellow
Form:
Liquid
Lyophilized
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type Color Form
23-666-479 1 x 2 mL Lipoprotein A Calibrator Set Light Yellow Lyophilized
23-666-140 1 x 2 mL AutoHDL™/LDL Calibrator Light Yellow Lyophilized
23-666-371 6 x 1 mL Microalbumin Calibrator Set Clear Liquid
23-666-189 10 x 5 mL Chemistry Calibrator Light Yellow Lyophilized
23-666-203 - - - -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 23-666-479 Supplier Pointe Scientific Supplier No. L7597CAL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Liquid-stable, serum-based calibrator sets contain both Total and Direct Bilirubin values

•    Chemistry Calibrator (23-666-189): human serum-based (lyophilized)
•    Cholesterol Calibrator (23-666-203): human serum-based (lyophilized) with cholesterol value of approximately 50mg/dL

Specifications

Certifications/Compliance CE
Color Light Yellow
Content And Storage 2°C to 8°C
Type Lipoprotein A Calibrator Set
Includes Sodium Azide <0.1%
Intended Use In-vitro diagnostic use only
Quantity 1 x 2 mL
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.