Learn More
- Home
- Products
- Research and Clinical Diagnostic Tests and Controls
- Chemistry Controls, Calibrators and Reagents
- Clinical Chemistry Standards
- Pointe Scientific General Chemistry Standards
Pointe Scientific General Chemistry Standards
Catalog No. 23666304 Shop All Pointe Scientific Products| Catalog No. | Quantity | Type | Concentration |
|---|---|---|---|
| 23-666-304 | 1 x 2 mL | Hemoglobin Standard | 12 g/dL |
| 23-666-356 | 1 x 3 mL | Lipase Standard Reagent Set | 600 U/L |
| 23-666-424 | 1 x 15 mL | Uric Acid Standard | 0.005 g/dL |
| 23-666-199 | 1 x 15 mL | Cholesterol - HDL Standard | 0.05 g/dL |
| 23-666-341 | 1 x 15 mL | Lactate Standard | 5 mM/L |
| 23-666-360 | 1 x 15 mL | Magnesium Standard Reagent Set | 0.001 to 0.002 g/dL |
| 23-666-101 | 1 x 4 mL | Ammonia Standard | 500 μm/L |
| 23-666-277 | 1 x 15 mL | Glucose Standard | 0.1 g/dL |
| 23-666-182 | 1 x 15 mL | Carbon Dioxide Standard | 30 mM/L |
| 23-666-312 | 1 x 30 mL | Iron Standard | 500 μg/dL |
| 23-666-167 | 1 x 15 mL | Calcium Standard | 0.01 g/dL |
| 23-666-100 | 1 x 4 mL | Ammonia Standard | 100 μm/L |
| 23-666-071 | 1 x 5 mL | Alcohol Standard | 0.1 g/dL |
| 23-666-373 | 1 x 15 mL | Microprotein Standard Set | 0.05 g/dL |
| 23-666-153 | 1 x 15 mL | BUN Standard | 0.02 g/dL |
| 23-666-235 | 1 x 15 mL | Creatinine Standard | 0.002 g/dL |
| 23-666-422 | 1 x 15 mL | Triglyceride Standard | 0.2 g/dL |
| 23-666-388 | 1 x 15 mL | Total Protein Standard Set | 8 g/dL |
| 23-666-198 | 1 x 15 mL | Cholesterol Standard | 0.2 g/dL |
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area.
Specifications
Specifications
| Certifications/Compliance | CE |
| Color | Dark Red |
| Content And Storage | 2°C to 8°C |
| Concentration | 12 g/dL |
| Type | Hemoglobin Standard |
| Includes | Human Blood cells |
| Intended Use | For use as Human hemoglobin standard for the cyanmethemoglobin hemoglobin procedure |
| Quantity | 1 x 2 mL |
| Stability | 1 Month |
| Form | Lyophilized |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.