Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific General Chemistry Standards

Catalog No. 23666424 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
1 x 2 mL
1 x 3 mL
1 x 4 mL
1 x 5 mL
1 x 15 mL
1 x 30 mL
Type:
Alcohol Standard
Ammonia Standard
BUN Standard
Calcium Standard
Carbon Dioxide Standard
Cholesterol - HDL Standard
Cholesterol Standard
Creatinine Standard
Glucose Standard
Hemoglobin Standard
Iron Standard
Lactate Standard
Lipase Standard Reagent Set
Magnesium Standard Reagent Set
Microprotein Standard Set
Total Protein Standard Set
Triglyceride Standard
Uric Acid Standard
Concentration:
0.001 to 0.002 g/dL
0.002 g/dL
0.005 g/dL
0.01 g/dL
0.02 g/dL
0.05 g/dL
0.1 g/dL
0.2 g/dL
100 μm/L
12 g/dL
30 mM/L
5 mM/L
500 μg/dL
500 μm/L
600 U/L
8 g/dL
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type Concentration
23-666-424 1 x 15 mL Uric Acid Standard 0.005 g/dL
23-666-356 1 x 3 mL Lipase Standard Reagent Set 600 U/L
23-666-199 1 x 15 mL Cholesterol - HDL Standard 0.05 g/dL
23-666-341 1 x 15 mL Lactate Standard 5 mM/L
23-666-360 1 x 15 mL Magnesium Standard Reagent Set 0.001 to 0.002 g/dL
23-666-101 1 x 4 mL Ammonia Standard 500 μm/L
23-666-277 1 x 15 mL Glucose Standard 0.1 g/dL
23-666-182 1 x 15 mL Carbon Dioxide Standard 30 mM/L
23-666-312 1 x 30 mL Iron Standard 500 μg/dL
23-666-167 1 x 15 mL Calcium Standard 0.01 g/dL
23-666-100 1 x 4 mL Ammonia Standard 100 μm/L
23-666-071 1 x 5 mL Alcohol Standard 0.1 g/dL
23-666-373 1 x 15 mL Microprotein Standard Set 0.05 g/dL
23-666-153 1 x 15 mL BUN Standard 0.02 g/dL
23-666-235 1 x 15 mL Creatinine Standard 0.002 g/dL
23-666-422 1 x 15 mL Triglyceride Standard 0.2 g/dL
23-666-388 1 x 15 mL Total Protein Standard Set 8 g/dL
23-666-198 1 x 15 mL Cholesterol Standard 0.2 g/dL
23-666-304 1 x 2 mL Hemoglobin Standard 12 g/dL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
This item is not returnable. View return policy
Catalog No. 23-666-424 Supplier Pointe Scientific Supplier No. U7580STD
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Certifications/Compliance CE
Color Clear
Content And Storage 2°C to 8°C
Concentration 0.005 g/dL
Type Uric Acid Standard
Includes Sodium Azide 0.05%
Quantity 1 x 15 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.