Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quanterix Inc GENERAL DETECTOR/SAMPLE DILUEN
SDP

Catalog No. NC0851965
Encompass

  • Designed for sensitive immunoassay applications.
  • Facilitates detection and quantification of purified proteins.
  • Provides a stable, optimized environment for research-grade immunoassays.
  • Formulation includes proteins, surfactants, preservatives, and interference blockers.
  • Intended for research use only; not for diagnostic purposes.
  • Compatible with Quanterix Simoa HD-1, HD-X, and SR-X instruments.
  • Water-based buffer base.
  • Volume of 250 mL.
  • Storage conditions of 2-8°C.
  • Hazard classification: Not classified (GHS-US).
  • Safety profile: Non-hazardous, non-toxic, non-infectious.

Catalog No. NC0851965 Supplier Quanterix Inc Supplier No. 101359
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Designed for sensitive immunoassay applications.
  • Facilitates detection and quantification of purified proteins.
  • Provides a stable, optimized environment for research-grade immunoassays.
  • Formulation includes proteins, surfactants, preservatives, and interference blockers.
  • Intended for research use only; not for diagnostic purposes.
  • Compatible with Quanterix Simoa HD-1, HD-X, and SR-X instruments.
  • Water-based buffer base.
  • Volume of 250 mL.
  • Storage conditions of 2-8°C.
  • Hazard classification: Not classified (GHS-US).
  • Safety profile: Non-hazardous, non-toxic, non-infectious.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.