Learn More
MSC General DM8200 Handheld Digital Manometer, 100 Max PSI, 1 ea
Catalog No. 5000506717
Encompass
This handheld digital manometer offers precise pressure measurement with a maximum capacity of 100 psi and 0.3% accuracy, capable of measuring up to 2768 Inch Water Column. It is designed for various laboratory and industrial applications requiring accurate pressure readings.
  • LCD display for clear readings
  • Durable plastic case for protection
  • Wide operating temperature range from 0 °C (32 °F)
  • Includes a custom hard plastic carrying case, two connection hoses, and a battery
  • Complies with Prop 65 standards
Catalog No. 50-005-06717 Supplier MSC Supplier No. 74451071
This handheld digital manometer offers precise pressure measurement with a maximum capacity of 100 psi and 0.3% accuracy, capable of measuring up to 2768 Inch Water Column. It is designed for various laboratory and industrial applications requiring accurate pressure readings.
  • LCD display for clear readings
  • Durable plastic case for protection
  • Wide operating temperature range from 0 °C (32 °F)
  • Includes a custom hard plastic carrying case, two connection hoses, and a battery
  • Complies with Prop 65 standards