Learn More
Braintree Scientific General Lab Animal Tattoo System, Quiet System
SDP
Catalog No. 501954220
Encompass
Convenient comprehensive kits for the humane tattoo identification of neonate or adult mice and rats. Each kit is provided with a comprehensive training guide and instruction DVD and all the tools needed to safely and efficiently tattoo rodents. AIMS tattooing techniques are performed on unanesthetized animals and produce easy-to-read permanent ID's. AIMS tattoo pigments and supplies have been used in the industry for over 25 years and have been demonstrated to be safe and non-toxic. Tattoo identification is cost effective with an average disposable cost of about 2 per neonate and 5 per adult rodent. Same Specifications and Supplies as the original systems this Quiet Tattoo Machine version Reduces noise level which leads to a calmer animal, 54% lighter, better balance, less hand fatigue. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4220 Supplier Braintree Scientific Supplier No. ATS3Q
Convenient comprehensive kits for the humane tattoo identification of neonate or adult mice and rats. Each kit is provided with a comprehensive training guide and instruction DVD and all the tools needed to safely and efficiently tattoo rodents. AIMS tattooing techniques are performed on unanesthetized animals and produce easy-to-read permanent ID's. AIMS tattoo pigments and supplies have been used in the industry for over 25 years and have been demonstrated to be safe and non-toxic. Tattoo identification is cost effective with an average disposable cost of about 2 per neonate and 5 per adult rodent. Same Specifications and Supplies as the original systems this Quiet Tattoo Machine version Reduces noise level which leads to a calmer animal, 54% lighter, better balance, less hand fatigue. **Sold for research purposes only and not for human use**