Learn More
3B Scientific™ General Parasitology, Short Set German Slides
SDPSCI_ED
Catalog No. S00555 Shop All 3B Scientific Products
Change view
Click to view available options
Includes:
25 Microscope Slides
Catalog No. Includes
S00555 25 Microscope Slides
1 options
This item is not returnable. View return policy
Catalog No. S00555 Supplier 3B Scientific™ Supplier No. 1004214
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
General Parasitology, Short Set German Slides includes 25 Microscope Slides.
  • 25 Microscope Slides.
    • 1(f) Trypanosoma gambiense, Central African sleeping disease, blood smear
    • 2(f) Plasmodium berghei, malaria in rodents, blood smear with vegetative forms and schizogony stages
    • 3(f) Sarcocystis sp., section of muscle showing the parasites in Miescher's tubes
    • 4(e) Nosema apis, honey bee dysentery, t.s. of diseased bee intestine
    • 5(d) Eimeria stiedae, causes coocidiosis in rabbit liver, t.s. shows parasites in all stages
    • 6(f) Fasciola hepatica, beef liver fluke, w.m. of adult flat mount and carefully stained
    • 7(d) Fasciola hepatica, ova w.m.
    • 8(t) Taenia or Moniezia, tapeworm, scolex w.m.
    • 9(f) Taenia pisiformis, dog tapeworm, mature proglottids w.m.
    • 10(d) Taenia saginata, tapeworm, proglottids in different stages t.s.
    • 11(f) Hymenolepis nana, dwarf tapeworm, proglottids w.m.
    • 12(f) Echinococcus granulosus, cyst wall and scolices sec.
    • 13(d) Ascaris lumbricoides, roundworm of human, adult female t.s. in region of gonads
    • 14(d) Ascaris lumbricoides, ova from faeces w.m.
    • 15(f) Enterobius vermicularis (Oxyuris), pin worm, adult specimen w.m.
    • 16(d) Trichinella spiralis, muscle with encysted larvae l.s.
    • 17(g) Ixodes sp., tick, adult w.m. Carrier of relapsing fever and borreliosis
    • 18(d) Dermanyssus gallinae, chicken mite w.m.
    • 19(e) Acarapis woodi, varroa, parasitic mite of honey bee, w.m.
    • 20(e) Sarcoptes scabiei (Acarus siro), section of diseased skin with parasites
    • 21(f) Anopheles, malaria mosquito, head and mouth parts of female w.m.
    • 22(e) Culex pipiens, common mosquito, head and mouth parts of female w.m.
    • 23(f) Cimex lectularius, bed bug, w.m.
    • 24(f) Pediculus humanus, human louse, w.m.
    • 25(e) Ctenocephalus canis, dog flea, adult w.m.
Specifications
Product Type Parasitology Slides
Includes 25 Microscope Slides