Learn More
RWD LIFE SCIENCE INC GENERAL PROBE HOLDER 6.0MM-12
SDP
Catalog No. NC2748758
Encompass
Lateral fixation, holds micro syringe needles, large diameter electrodes and flexible drill handles etc, V-groove design with 6mm-12mm diameters
Catalog No. NC2748758 Supplier RWD LIFE SCIENCE INC Supplier No. 68202
Lateral fixation, holds micro syringe needles, large diameter electrodes and flexible drill handles etc, V-groove design with 6mm-12mm diameters