Learn More
Electron Microscopy Sciences General Protocol Incubator Model 51028063, 120 V, 75 L chamber
SDP
Catalog No. 501902387
Encompass_Preferred
Gravity convection provides gentle air flow and minimal drying out with temperatures at or below ambient: +5 to 75 degree C. The intuitive user interface for setting temperature and large and easy to read vacuum fluorescent display makes operation simple and straight forward operation. Internal glass door allows sample viewing without impacting temperature. Easy to clean, corrosion-resistant stainless-steel interior (AISI 430) a plus.
Chamber: 75 L
Catalog No. 50-190-2387 Supplier Electron Microscopy Sciences Supplier No. 6315310
Gravity convection provides gentle air flow and minimal drying out with temperatures at or below ambient: +5 to 75 degree C. The intuitive user interface for setting temperature and large and easy to read vacuum fluorescent display makes operation simple and straight forward operation. Internal glass door allows sample viewing without impacting temperature. Easy to clean, corrosion-resistant stainless-steel interior (AISI 430) a plus.
Chamber: 75 L