Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Safety-Walk™ General Purpose Cleats, Series 600

Catalog No. 17623934 Shop All 3M Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-623-934 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-623-934 Supplier 3M™ Safety-Walk™ Supplier No. 7100122465
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

This mineral coated, slip resistant material features a highly durable surface. Low profile design helps reduce trip hazards.

  • Anti-slip tape for light to heavy shoe-traffic areas
  • High quality abrasive particles ensure long-lasting friction in any application
  • Highly durable surface withstands the rigors of equipment traffic
  • Low-profile design helps reduce tripping hazards
  • High performance, pressure-sensitive adhesive for easy application
  • NFSI High Traction Certification
TRUSTED_SUSTAINABILITY

Specifications

Finish Mineral-coated
Length (Metric) 610 mm
Packaging Quantity 50 Pk.
Type Floor Tape
Dimensions (L x W) 610 x 19.05 mm
Width (English) 0.75 in.
Width (Metric) 19.05 mm
For Use With (Application) Light to Heavy Traffic Areas
Length (English) 24 in.
Adhesive Type Pressure Sensitive Adhesive
Color Black
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.