Learn More
Andwin Scientific GENERAL PURPOSE LASER TOUGH-TA
SDP
Catalog No. 50820962
Encompass_Preferred
The General Purpose Tough-Tags feature resistance against chemicals and capacity to work within the temperature range of -40C to 121C without peeling, cracking, or falling off. The labels will not jam in laser printers. This product comes with guidelines for printing with Microsoft Word. The labels strongly stick onto several surfaces like plastics and other materials and measure 1.5" x 0.75".
Catalog No. 50-820-962 Supplier Andwin Scientific Supplier No. 91471050
The General Purpose Tough-Tags feature resistance against chemicals and capacity to work within the temperature range of -40C to 121C without peeling, cracking, or falling off. The labels will not jam in laser printers. This product comes with guidelines for printing with Microsoft Word. The labels strongly stick onto several surfaces like plastics and other materials and measure 1.5" x 0.75".