Learn More
Felix Storch Inc ACCUCOLD General Purpose Medical Freezer
SDP
Catalog No. NC2250135
Encompass_Preferred
The VT65MLGP is a counter height all-refrigerator designed to offer an improved performance over similarly sized dorm fridges and residential units. This model has a white cabinet and user-reversible door, with a factory-installed lock for added security. A digital thermostat makes temperature management easy and efficient. This unit is designed for freestanding use. Inside, the 24" wide all freezers has a low temperature manual defrost operation that can reach -25ºC. Three slide-out basket drawers offer convenient storage that keeps items cold when the door is opened. A one-piece floor liner makes cleanup easy: https://www.accucold.com/product/VT65MLGP
This item is not returnable. View return policy
Catalog No. NC2250135 Supplier Felix Storch Inc Supplier No. VT65MLGP
The VT65MLGP is a counter height all-refrigerator designed to offer an improved performance over similarly sized dorm fridges and residential units. This model has a white cabinet and user-reversible door, with a factory-installed lock for added security. A digital thermostat makes temperature management easy and efficient. This unit is designed for freestanding use. Inside, the 24" wide all freezers has a low temperature manual defrost operation that can reach -25ºC. Three slide-out basket drawers offer convenient storage that keeps items cold when the door is opened. A one-piece floor liner makes cleanup easy: https://www.accucold.com/product/VT65MLGP