Learn More
Perkin Elmer US LLC General Purpose Optical Bench - Extension Pack
Catalog No. 502113699
Encompass_Preferred
This pack requires, but does not include either the General Purpose Optical Bench with the MCT detector (L1250425) or the General Purpose Optical Bench with the L1TaO3 detector (l1250463).
Catalog No. 50-211-3699 Supplier Perkin Elmer US LLC Supplier No. L1272501
This pack requires, but does not include either the General Purpose Optical Bench with the MCT detector (L1250425) or the General Purpose Optical Bench with the L1TaO3 detector (l1250463).