Learn More
Marketlab General Purpose Probe Cover, Sterile
Catalog No. 22505281
Change view
Click to view available options
Length (English):
24 in.
36 in.
58 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
22-505-281 24 in.
22-505-165 36 in.
22-505-252 58 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-505-281 Supplier Marketlab Supplier No. 914609
Add to Cart
Add to Cart
Contains a specially formulated material with scanning properties similar to human tissue.
  • Designed specifically for ultrasound use to easily slide over probes and provide distortion-free imaging.
  • These covers are a recommended alternative for latex-sensitive patients and professionals.
  • Made popular for OB/GYN and sterile scanning procedures.
  • Soft, stretchable material is comfortable for patient
  • Each box is conveniently packaged with sterile gel and elastic bands for 24 procedures
  • Contains a specially formulated material with scanning properties similar to human tissue
  • Cover with box-style 3D-end helps provide better probe coupling
Specifications
Quantity 24/Pk.
Length (English) 24 in.