Learn More
Alkali Scientific ASI™ General Purpose Sealing Film, 100/pk, Sterile
SDP
Catalog No. 502388708
Encompass_Preferred
General Purpose polypropylene films offer superior barrier properties to minimize evaporation and prevent well-to-well contamination. Ideal for use with polypropylene plates.Certified DNase- & RNase- free. Features good chemical resistance. Provides a very secure seal to minimize evaporation, but is slightly harder to remove than SealPlate films. Less rigid polypropylene film reduces aerosols upon removal. Specially formulated adhesive minimizes plate residue. Can be used as a temporary seal while awaiting further processing.
Catalog No. 50-238-8708 Supplier Alkali Scientific Supplier No. SF18MLS
General Purpose polypropylene films offer superior barrier properties to minimize evaporation and prevent well-to-well contamination. Ideal for use with polypropylene plates.Certified DNase- & RNase- free. Features good chemical resistance. Provides a very secure seal to minimize evaporation, but is slightly harder to remove than SealPlate films. Less rigid polypropylene film reduces aerosols upon removal. Specially formulated adhesive minimizes plate residue. Can be used as a temporary seal while awaiting further processing.