Learn More
Surgical Design™ General-Purpose Tooth Luxation Elevator, No. 301
Catalog No. 21124704 Shop All Surgical Design Products
Change view
Click to view available options
Size:
Adult
Catalog No. Size
21-124-704 Adult
1 options
Catalog No. 21-124-704 Supplier Surgical Design™ Supplier No. DELAPX301
Add to Cart
Add to Cart
No. 301 tooth luxation elevator supports controlled periodontal ligament separation and atraumatic tooth elevation for general dentistry and extraction prep.
  • Model: No. 301 General-Purpose Luxation Elevator
  • Designed for precise periodontal ligament severing prior to extraction
  • Slim, tapered blade allows controlled insertion along the root surface
  • Facilitates atraumatic elevation, reducing risk of root or bone damage
  • Ergonomic, textured handle provides secure, slip-resistant grip
  • Balanced weight for improved maneuverability and tactile feedback
  • Crafted from high-quality surgical stainless steel
  • Autoclavable and reusable for long-term clinical use
  • Suitable for general dentistry, oral surgery, and emergency dental procedures
Specifications
Product Type Apexo Elevator
Handle Material Stainless Steel
Length (Metric) 15.5 cm
Material Stainless Steel
Size Adult
Autoclavable Autoclavable
Color Silver