Learn More
Checkers General-Purpose Utility Step Wheel Chock with Cored Bottom
GREENER_CHOICE
Catalog No. 19088420 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Orange
Catalog No. Color
19-088-420 Orange
1 options
Catalog No. 19-088-420 Supplier Checkers Supplier No. UCTS003
Add to Cart
Add to Cart
Checkers heavy-duty UC series wheel chock is manufactured with highly durable, lightweight polyurethane construction.
  • Durable polyurethane construction increases strength over comparable rubber chocks
  • Resists damage from UV, moisture, salt, and oils/chemicals
  • Grooved, stepped edges help grip and provide more tire traction
  • Eyebolt to use as a handle or connect with a rope
  • Up to a 20,000 lb. load capacity (up to 11% grade)
Specifications
Material Urethane
Product Type Wheel Chock
Color Orange