Learn More
Checkers General-Purpose Utility Wheel Chock
GREENER_CHOICE
Catalog No. 17456119 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Orange
Catalog No. Color
17-456-119 Orange
1 options
Catalog No. 17-456-119 Supplier Checkers Supplier No. UC1600
Add to Cart
Add to Cart
General-Purpose Utility Wheel Chock.
  • For tires with up to a 35 in. outside dia.
  • Rated for vehicles with a gross operating weight up to 40,000 lbs.
  • Weather-resistant, lightweight polyurethane construction
  • Resistant to oils, fuels, and solvents
  • Recessed carrying handle
  • Mounting hole for ropes or chains
  • High-visibility orange color
  • Available in singles or pairs, specify after model number S=Single or P=Pair
Specifications
Material Polyurethane
Product Type Wheel Chock
Color Orange