Learn More
Brady™ General Purpose Vinyl Labels
Catalog No. 17108837 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
1.90 x 3 in.
Catalog No. Dimensions (L x W)
17-108-837 1.90 x 3 in.
1 options
Catalog No. 17-108-837 Supplier Brady™ Supplier No. M7377569CLP4B
Add to Cart
Add to Cart
This flexible vinyl film (B-7569) with ultra-aggressive adhesive sticks permanently to rough, highly textured and difficult surfaces.
  • Permanent acrylic adhesive sticks to a wide variety of surfaces.
  • Vinyl film withstands exposure to moisture, chemicals and handling both indoor/outdoor, with an average outdoor lifespan up to three years
  • A ribbon is required to print this material - use part number M7-R10000
Specifications
Adhesive Type Acrylic
Color Red on White
Finish Gloss
Format Label
Material Vinyl
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
Dimensions (L x W) 1.90 x 3 in.
Width (English) 3 in.