Learn More
Sigma Aldrich Fine Chemicals Biosciences General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified viscosity 258 mPa.s (25 C) | 500ML
Catalog No. 501765408
Encompass
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified viscosity 258 mPa.s (25 C) | 500ML
Catalog No. 50-176-5408 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. N140500ML
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified viscosity 258 mPa.s (25 C) | 500ML