Learn More
Chemglass Life Sciences Threaded Stopcock, General Purpose, 90°, Complete
SDP
Catalog No. CG58701
Encompass_Preferred
Threaded Stopcock, General Purpose, 90¿, Complete. A 2mm bore grease-free, general purpose stopcock is designed primarily for use in control of fluids. Excellent for use in construction of burettes, addition funnels and non-vacuum apparatus. Valve is constructed so that only glass and PTFE come in contact with fluids. Tip design provides for semi-needle valve metering action. Sidearms are 8mm OD and sealed to barrel at a 90¿ angle to one another.
Catalog No. CG58701 Supplier Chemglass Life Sciences Supplier No. CG58701
Threaded Stopcock, General Purpose, 90¿, Complete. A 2mm bore grease-free, general purpose stopcock is designed primarily for use in control of fluids. Excellent for use in construction of burettes, addition funnels and non-vacuum apparatus. Valve is constructed so that only glass and PTFE come in contact with fluids. Tip design provides for semi-needle valve metering action. Sidearms are 8mm OD and sealed to barrel at a 90¿ angle to one another.