Learn More
Cloud Clone Corp General species Estradiol (E2) (Competitive ELISA) ELISA Kit
Catalog No. NC4011093
Encompass_Preferred
General species Estradiol (E2) (Competitive ELISA) ELISA Kit measures General species E2 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids.This assay has high sensitivity and excellent specificity for detection of Estradiol (E2).No significant cross-reactivity or interference between Estradiol (E2) and analogues was observed.Detection Range: 12.35-1,000pg/mLSensitivity:The minimum detectable dose of this kit is typically less than 4.93pg/mLDetection menthod: ColormetricGene ID:-
Catalog No. NC4011093 Supplier Cloud Clone Corp Supplier No. CEA461GE
General species Estradiol (E2) (Competitive ELISA) ELISA Kit measures General species E2 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids.This assay has high sensitivity and excellent specificity for detection of Estradiol (E2).No significant cross-reactivity or interference between Estradiol (E2) and analogues was observed.Detection Range: 12.35-1,000pg/mLSensitivity:The minimum detectable dose of this kit is typically less than 4.93pg/mLDetection menthod: ColormetricGene ID:-