Learn More
Cloud Clone Corp General species Lipopolysaccharide (LPS) (Sandwich ELISA) ELISA Kit
Catalog No. NC4011163
Encompass_Preferred
General species Lipopolysaccharide (LPS) (Sandwich ELISA) ELISA Kit measures General species LPS in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids.This assay has high sensitivity and excellent specificity for detection of Lipopolysaccharide (LPS).No significant cross-reactivity or interference between Lipopolysaccharide (LPS) and analogues was observed.Detection Range: 0.47-30ng/mLSensitivity:The minimum detectable dose of this kit is typically less than 0.17ng/mLDetection menthod: ColormetricGene ID:-
Catalog No. NC4011163 Supplier Cloud Clone Corp Supplier No. SEB526GE
General species Lipopolysaccharide (LPS) (Sandwich ELISA) ELISA Kit measures General species LPS in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids.This assay has high sensitivity and excellent specificity for detection of Lipopolysaccharide (LPS).No significant cross-reactivity or interference between Lipopolysaccharide (LPS) and analogues was observed.Detection Range: 0.47-30ng/mLSensitivity:The minimum detectable dose of this kit is typically less than 0.17ng/mLDetection menthod: ColormetricGene ID:-