Learn More
Cloud Clone Corp General species Testosterone (Testo) (Competitive ELISA) ELISA Kit
Catalog No. NC4011110
Encompass_Preferred
General species Testosterone (Testo) (Competitive ELISA) ELISA Kit measures General species Testo in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids.This assay has high sensitivity and excellent specificity for detection of Testosterone (Testo).No significant cross-reactivity or interference between Testosterone (Testo) and analogues was observed.Detection Range: 370.4-30,000pg/mLSensitivity:The minimum detectable dose of this kit is typically less than 152.5pg/mLDetection menthod: ColormetricGene ID:-
Catalog No. NC4011110 Supplier Cloud Clone Corp Supplier No. CEA458GE
General species Testosterone (Testo) (Competitive ELISA) ELISA Kit measures General species Testo in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids.This assay has high sensitivity and excellent specificity for detection of Testosterone (Testo).No significant cross-reactivity or interference between Testosterone (Testo) and analogues was observed.Detection Range: 370.4-30,000pg/mLSensitivity:The minimum detectable dose of this kit is typically less than 152.5pg/mLDetection menthod: ColormetricGene ID:-