Learn More
MSC General Steel Rule, 6 in, 4R English & Metric, Rigid, 3/4 in
Catalog No. 5000527463
Encompass
The steel rule features a 6-inch overall length and a rigid stainless steel construction, making it suitable for precise measurements. It includes 4R English and metric graduations for versatile use in various applications.
  • Overall length: 6 in
  • Graduation: 4R English & metric, including 1/16, 1/32, 1/64 & 1/8 in
  • Rigid flexibility
  • Overall width: 3/4 in
  • Polished silver finish
  • Stainless steel material
  • Silver color
  • Does not include hook
  • Not magnetic
  • Horizontal scale orientation
Catalog No. 50-005-27463 Supplier MSC Supplier No. 56208010
Add to Cart
Add to Cart
The steel rule features a 6-inch overall length and a rigid stainless steel construction, making it suitable for precise measurements. It includes 4R English and metric graduations for versatile use in various applications.
  • Overall length: 6 in
  • Graduation: 4R English & metric, including 1/16, 1/32, 1/64 & 1/8 in
  • Rigid flexibility
  • Overall width: 3/4 in
  • Polished silver finish
  • Stainless steel material
  • Silver color
  • Does not include hook
  • Not magnetic
  • Horizontal scale orientation