Learn More
Med Vet International 19-pc. Economy General Surgery Pack
Catalog No. 502224146
Encompass_Preferred
The General Surgery Pack Includes (1) 6.75 in. Mayo Scissors, Straight (1) 7" Metzenbaum Scissors, Curved (1) 4.75 in. #3 Scalpel/Blade Handle (3) 5" Halstead Mosquito Forceps, Curved (2) 5.5 in. Kelly Hemostat Forceps, Curved (4) 6.25 in. Rochester-Carmalt Forceps, Curved (4) 3.5 in. Backhaus Towel Forceps (1) 6.5 in. Olsen-Hegar Needle Holder (1) 4.75 in. Adson-Brown Forceps, 7x7 Teeth (1) 8" Snook Hook
Catalog No. 50-222-4146 Supplier Med Vet International Supplier No. GPACKECON
The General Surgery Pack Includes (1) 6.75 in. Mayo Scissors, Straight (1) 7" Metzenbaum Scissors, Curved (1) 4.75 in. #3 Scalpel/Blade Handle (3) 5" Halstead Mosquito Forceps, Curved (2) 5.5 in. Kelly Hemostat Forceps, Curved (4) 6.25 in. Rochester-Carmalt Forceps, Curved (4) 3.5 in. Backhaus Towel Forceps (1) 6.5 in. Olsen-Hegar Needle Holder (1) 4.75 in. Adson-Brown Forceps, 7x7 Teeth (1) 8" Snook Hook