Learn More
Med Vet International 19-pc. General Surgery Pack, Von Klaus German Surgical Steel
Catalog No. 502092178
Encompass_Preferred
The Von Klaus line of instruments is made of high quality German surgical steel, expertly crafted and comes with a lifetime warranty when used for their intended purpose.Instruments Include (1) 6.75-in. Mayo Scissors, Straight (1) 7-in. Metzenbaum Scissors, Curved (1) 4.75-in. #3 Scalpel/Blade Handle (3) 5-in. Halstead Mosquito Forceps, Curved (2) 5.5-in. Kelly Hemostat Forceps, Curved (4) 6.25-in. Rochester-Carmalt Forceps, Curved (4) 3.5-in. Backhaus Towel Forceps (1) 6.5-in. Olsen-Hegar Needle Holder (1) 4.75-in. Adson-Brown Forceps, 7x7 Teeth (1) 8-in. Snook Hook
Catalog No. 50-209-2178 Supplier Med Vet International Supplier No. VKPACK1
The Von Klaus line of instruments is made of high quality German surgical steel, expertly crafted and comes with a lifetime warranty when used for their intended purpose.Instruments Include (1) 6.75-in. Mayo Scissors, Straight (1) 7-in. Metzenbaum Scissors, Curved (1) 4.75-in. #3 Scalpel/Blade Handle (3) 5-in. Halstead Mosquito Forceps, Curved (2) 5.5-in. Kelly Hemostat Forceps, Curved (4) 6.25-in. Rochester-Carmalt Forceps, Curved (4) 3.5-in. Backhaus Towel Forceps (1) 6.5-in. Olsen-Hegar Needle Holder (1) 4.75-in. Adson-Brown Forceps, 7x7 Teeth (1) 8-in. Snook Hook