Learn More
MSC General Steel Rule, 12 in. Length, 5R Graduation, Flexible, 1 ea.
Catalog No. 5000547882
Encompass
This flexible steel rule is a versatile tool designed for precise dimensional measurement. Crafted with a durable construction, it features a clear, satin chrome finish that reduces glare for improved readability during inspection and layout tasks.
  • Features 5R graduation style
  • Graduations provided in 32nd, 64th, 10th and 100th of an inch
  • Designed with ultra-fine black markings
  • Flexible material construction allows for measuring curved surfaces
  • Meets or exceeds U.S. Government Specification GGG-R-791f
  • Includes protective plastic sleeve
Catalog No. 50-005-47882 Supplier MSC Supplier No. 56207251
This flexible steel rule is a versatile tool designed for precise dimensional measurement. Crafted with a durable construction, it features a clear, satin chrome finish that reduces glare for improved readability during inspection and layout tasks.
  • Features 5R graduation style
  • Graduations provided in 32nd, 64th, 10th and 100th of an inch
  • Designed with ultra-fine black markings
  • Flexible material construction allows for measuring curved surfaces
  • Meets or exceeds U.S. Government Specification GGG-R-791f
  • Includes protective plastic sleeve