Learn More
MSC General Steel Rule, Flexible, 6 in., Decimal and Metric, 1 ea
Catalog No. 5000514517
Encompass
This flexible steel rule is designed for precise dimensional measurement, featuring both decimal and metric graduations to accommodate diverse project needs. It is finished with a hard satin chrome coating that reduces glare and includes ultra-fine black markings for high readability.
  • Graduations include every 10th and 100th of an inch, as well as mm and 0.5 mm increments
  • Flexible steel construction for versatility
  • Meets or exceeds U.S. Government Specification GGG-R-791f for accuracy
  • Hard satin chrome finish with ultra-fine black markings
  • Compact dimensions of 6 in. x 0.5 in.
  • Includes a plastic sleeve for protection
Catalog No. 50-005-14517 Supplier MSC Supplier No. 56207350
This flexible steel rule is designed for precise dimensional measurement, featuring both decimal and metric graduations to accommodate diverse project needs. It is finished with a hard satin chrome coating that reduces glare and includes ultra-fine black markings for high readability.
  • Graduations include every 10th and 100th of an inch, as well as mm and 0.5 mm increments
  • Flexible steel construction for versatility
  • Meets or exceeds U.S. Government Specification GGG-R-791f for accuracy
  • Hard satin chrome finish with ultra-fine black markings
  • Compact dimensions of 6 in. x 0.5 in.
  • Includes a plastic sleeve for protection