Learn More
MSC General Tools Steel Ruler, Steel, 12 in. / 300 mm, Flexible, Dual Graduations
Catalog No. 5000531127
Encompass
This flexible steel ruler is engineered for highly precise measurements, adhering to or surpassing U.S. Government Specification GGG-R-791f for accuracy. It features ultra-fine black graduations and is protected by a hard satin chrome finish. Each ruler comes with a plastic sleeve for storage.
  • Made from steel
  • 12 in. / 300 mm length
  • Graduations in 32nd and 64th of an inch, and millimeters
  • Flexible design
  • Hard satin chrome finish
  • Meets or exceeds U.S. Government Specification GGG-R-791f for accuracy
  • Includes a plastic sleeve for storage
  • Dimensions: 12 x 1/2 in.
Catalog No. 50-005-31127 Supplier MSC Supplier No. 56207277
This flexible steel ruler is engineered for highly precise measurements, adhering to or surpassing U.S. Government Specification GGG-R-791f for accuracy. It features ultra-fine black graduations and is protected by a hard satin chrome finish. Each ruler comes with a plastic sleeve for storage.
  • Made from steel
  • 12 in. / 300 mm length
  • Graduations in 32nd and 64th of an inch, and millimeters
  • Flexible design
  • Hard satin chrome finish
  • Meets or exceeds U.S. Government Specification GGG-R-791f for accuracy
  • Includes a plastic sleeve for storage
  • Dimensions: 12 x 1/2 in.