Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NewPath Learning™ Generating Electricity 3D Model Making Kit
SCI_ED

Catalog No. S436055 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
6 to 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S436055 6 to 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S436055 Supplier NewPath Learning™ Supplier No. 247950
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use this Generating Electricity 3D Model Making Kit, our interactive model of a power station, to teach your students how electricity is generated.

  • Teach your students how electricity is generated with our interactive model of a power station.
  • First, they will make their model using the colorful parts provided.
  • They will then use it to look at how electricity is generated from non-renewable fossil fuels or renewable bio-fuels.
  • The models covers different types of fuel, the role of the boiler, what a steam turbine does, and how the generator produces electricity.
  • They can even turn the turbine!
  • The model is a great starting point to consider different types of fuel and the pros and cons of generating electricity in different ways.
  • The Model Making Kit includes enough materials for students to assemble up to 5 paper models working cooperatively.
  • The comprehensive Teacher Resource Guide included provides illustrated readings, activities, key vocabulary and an assessment, along with teaching opportunities for students.

Specifications

For Use With (Application) Model Assembly
Grade 6 to 10
Includes 5 Models, Teacher Resource Guide, Vinyl Pouch
Length (English) 12 in.
Material Paper
Weight (English) 2 lb.
Width (English) 9 in.
Product Type 3D Model
Grade Level Intermediate - Middle School, High School
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.