Learn More
MSC Liphatech Rodenticide, Pellet, Place Packs, 100 x 25 gm Pail
Catalog No. 502837960
Encompass
Generation Pellet Place Packs are pre-measured rodenticide designed with a special pack film that emits an attractive aroma to rodents. The innovative packaging keeps the pellets fresh and prevents user exposure, making them ideal for hard-to-reach baiting areas and locations requiring protection from contamination or moisture.
  • Pre-measured place packs for easy deployment.
  • Special pack film emits an attractive aroma.
  • Packaging prevents user exposure to the rodenticide.
  • Keeps pellets fresh until opened by rodents.
  • Suitable for hard-to-reach baiting areas.
  • Effective in locations needing protection from contamination or moisture.
  • Contains Difethialone as the active ingredient.
  • Controls rats and mice.
  • Approved for indoor and outdoor use.
Catalog No. 50-283-7960 Supplier MSC Supplier No. 74314998
Generation Pellet Place Packs are pre-measured rodenticide designed with a special pack film that emits an attractive aroma to rodents. The innovative packaging keeps the pellets fresh and prevents user exposure, making them ideal for hard-to-reach baiting areas and locations requiring protection from contamination or moisture.
  • Pre-measured place packs for easy deployment.
  • Special pack film emits an attractive aroma.
  • Packaging prevents user exposure to the rodenticide.
  • Keeps pellets fresh until opened by rodents.
  • Suitable for hard-to-reach baiting areas.
  • Effective in locations needing protection from contamination or moisture.
  • Contains Difethialone as the active ingredient.
  • Controls rats and mice.
  • Approved for indoor and outdoor use.