Learn More
Cole-Parmer IKA 0594000 Homogenizer Rotor-Stator Generator, Coarse, 105mm x 10mm, 1-100 mL
Catalog No. NC2038664 Shop All Cole-Parmer Products
Encompass
Select a generator according to your sample volume, working solution, and needed dispersion type. The standard-capacity generator is available in three volume ranges. The large-capacity generator is available in both coarse (20 ìm) and fine (5 ìm) dispersion.
Catalog No. NC2038664 Supplier Cole-Parmer Supplier No. 0472010
Select a generator according to your sample volume, working solution, and needed dispersion type. The standard-capacity generator is available in three volume ranges. The large-capacity generator is available in both coarse (20 ìm) and fine (5 ìm) dispersion.